Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hyou1
Entrez Gene ID
12282
Gene Name
hypoxia up-regulated 1
Gene Aliases
AI415631, CBP-140, Cab140, Grp170, Orp150
Chromosome Location
Chr.9: 44379490 - 44392369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021395.4 NP_067370.3 9 - 10 1104 61
GenBank mRNA AF228709.1 - 8 - 9 991 61
AK128937.1 - 9 - 10 1051 61
AK145857.1 - 9 - 10 1052 61
AK155721.1 - 9 - 10 1052 61
AK157949.1 - 9 - 10 1022 61
AK171769.1 - 9 - 10 1052 61
BC019785.1 - 8 - 9 1010 61
BC050107.1 - 8 - 9 1009 61
BC058255.1 - 7 - 8 1302 61

View Full Product Details