The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031380.2 | NP_113557.1 | 4 - 5 | 758 | 73 |
GenBank mRNA | AB024429.1 | - | 4 - 5 | 731 | 73 |
AF276238.1 | - | 4 - 5 | 737 | 73 | |
AF310616.1 | - | 4 - 5 | 731 | 73 | |
AK079286.1 | - | 2 - 3 | 378 | 73 | |
AK082898.1 | - | 4 - 5 | 760 | 73 | |
BC061052.1 | - | 4 - 5 | 758 | 73 |