The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181816.2 | NP_861537.2 | 11 - 12 | 1501 | 66 |
GenBank mRNA | AK077147.1 | - | 6 - 7 | 879 | 66 |
BC096547.1 | - | 10 - 11 | 1394 | 66 | |
KC211186.1 | - | 11 - 12 | 1432 | 66 |