Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc67
Entrez Gene ID
234964
Gene Name
coiled-coil domain containing 67
Gene Aliases
4933401K09Rik, Deup1
Chromosome Location
Chr.9: 15559864 - 15628338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181816.2 NP_861537.2 1 - 2 107 125
GenBank mRNA AK030153.1 - 1 - 2 90 125
BY069891.1 - 1 - 2 108 125

View Full Product Details