The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113323.1 | NP_001106794.1 | 8 - 9 | 1026 | 134 |
GenBank mRNA | AK014852.1 | - | 8 - 9 | 859 | 134 |
BC132200.1 | - | 8 - 9 | 819 | 134 | |
BC132202.1 | - | 8 - 9 | 819 | 134 |