Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snn
Entrez Gene ID
20621
Gene Name
stannin
Gene Aliases
2810407J07Rik, AI848521, AW547286
Chromosome Location
Chr.16: 11066298 - 11074985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009223.3 NP_033249.1 1 - 2 150 79
GenBank mRNA AK149685.1 - 1 - 2 99 79
BC006961.1 - 1 - 2 118 79
BC017685.1 - 1 - 2 67 79
BC063027.1 - 1 - 2 87 79
CJ166486.1 - 1 - 2 102 79

View Full Product Details