Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Paqr5
Entrez Gene ID
74090
Gene Name
progestin and adipoQ receptor family member V
Gene Aliases
0610010I15Rik, AV002411, BB115488, mPRg
Chromosome Location
Chr.9: 61953738 - 62026790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028748.2 NP_083024.1 3 - 4 377 65
GenBank mRNA AK002481.1 - 3 - 4 347 65
AK035475.1 - 3 - 4 389 65
AK085411.1 - 3 - 4 378 65
AK086679.1 - 3 - 4 391 65
AY424294.1 - 3 - 4 347 65
BC054855.1 - 3 - 4 403 65
BC100516.1 - 2 - 3 258 65

View Full Product Details