Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfi2
Entrez Gene ID
30060
Gene Name
antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5
Gene Aliases
CD228, MTf
Chromosome Location
Chr.16: 31878501 - 31899020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013900.2 NP_038928.1 15 - 16 2258 143
GenBank mRNA AB024336.1 - 15 - 16 2258 143
AK036444.1 - 15 - 16 2193 143
BC040347.1 - 15 - 16 2181 143

View Full Product Details