Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Med15
Entrez Gene ID
94112
Gene Name
mediator complex subunit 15
Gene Aliases
A230074L19Rik, AW536074, Pcqap, mPcqap
Chromosome Location
Chr.16: 17651208 - 17722947 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040683.2 NP_001035773.1 4 - 5 368 133
NM_001285884.1 NP_001272813.1 4 - 5 368 133
NM_033609.3 NP_291087.2 4 - 5 368 133
GenBank mRNA AF328770.1 - 4 - 5 244 133
AK038915.1 - 4 - 5 331 133
AK131186.1 - 4 - 5 309 133
AK142224.1 - 4 - 5 334 133
AK169904.1 - 4 - 5 369 133
BC054779.1 - 4 - 5 318 133

View Full Product Details