Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Comt
Entrez Gene ID
12846
Gene Name
catechol-O-methyltransferase
Gene Aliases
Comt1, D16Wsu103e, D330014B15Rik
Chromosome Location
Chr.16: 18406882 - 18426716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111062.1 NP_001104532.1 2 - 3 413 82
GenBank mRNA AK148311.1 - 2 - 3 80 82

View Full Product Details