Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lctl
Entrez Gene ID
235435
Gene Name
lactase-like
Gene Aliases
E130104I05Rik, KLPH
Chromosome Location
Chr.9: 64117123 - 64138118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145835.2 NP_665834.1 1 - 2 158 59
GenBank mRNA AF309072.1 - 1 - 2 159 59
AK053515.1 - 1 - 2 47 59

View Full Product Details