Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam81a
Entrez Gene ID
76886
Gene Name
family with sequence similarity 81, member A
Gene Aliases
6430514L14Rik
Chromosome Location
Chr.9: 70089167 - 70142555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029784.2 NP_084060.1 1 - 2 243 81
GenBank mRNA AK018282.1 - 1 - 2 220 81
AK038532.1 - 1 - 2 200 81
AK044090.1 - 1 - 2 243 81
AK046408.1 - 1 - 2 136 81
AK079541.1 - 1 - 2 200 81
BC025646.1 - 1 - 2 269 81

View Full Product Details