The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081395.1 | NP_001074864.1 | 12 - 13 | 3020 | 66 |
GenBank mRNA | AK016526.1 | - | 13 - 14 | 2880 | 66 |
AK220238.1 | - | 10 - 11 | 2254 | 66 | |
AX763684.1 | - | 10 - 11 | 2544 | 66 | |
BC055117.1 | - | 4 - 5 | 745 | 66 | |
BC151000.1 | - | 12 - 13 | 2828 | 66 | |
BC151004.1 | - | 12 - 13 | 2828 | 66 | |
BC151007.1 | - | 12 - 13 | 2828 | 66 |