Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amotl1
Entrez Gene ID
75723
Gene Name
angiomotin-like 1
Gene Aliases
2310010G08Rik, 2310067L22Rik, 4932416D09Rik, JEAP, mFLJ00155
Chromosome Location
Chr.9: 14541966 - 14662960 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081395.1 NP_001074864.1 4 - 5 1660 69
GenBank mRNA AK016526.1 - 5 - 6 1520 69
AK220238.1 - 2 - 3 894 69
AX763684.1 - 2 - 3 1184 69
BC151000.1 - 4 - 5 1468 69
BC151004.1 - 4 - 5 1468 69
BC151007.1 - 4 - 5 1468 69

View Full Product Details