The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172925.2 | NP_766513.2 | 2 - 3 | 1264 | 62 |
GenBank mRNA | AK053085.1 | - | 2 - 3 | 1265 | 62 |
BC118027.1 | - | 2 - 3 | 1263 | 62 |