The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145977.1 | NP_001139449.1 | 2 - 3 | 888 | 100 |
NM_178721.4 | NP_848836.2 | 3 - 4 | 915 | 100 | |
GenBank mRNA | AK033973.1 | - | 2 - 3 | 515 | 100 |
AK038842.1 | - | 3 - 4 | 894 | 100 | |
AK043760.1 | - | 3 - 4 | 898 | 100 | |
AK046800.1 | - | 2 - 3 | 791 | 100 | |
AK163636.1 | - | 3 - 4 | 892 | 100 | |
AK165030.1 | - | 3 - 4 | 912 | 100 | |
BC139401.1 | - | 3 - 4 | 484 | 100 | |
BC139402.1 | - | 3 - 4 | 484 | 100 |