Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp60
Entrez Gene ID
22718
Gene Name
zinc finger protein 60
Gene Aliases
6330516O17Rik, AI426106, AI449354, Mfg-3, Mfg3
Chromosome Location
Chr.7: 27731409 - 27751689 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009560.2 NP_033586.2 1 - 2 262 70
GenBank mRNA AK028717.1 - 1 - 2 162 70
AK044869.1 - 1 - 2 261 70
CB526626.1 - 1 - 2 205 70

View Full Product Details