Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lss
Entrez Gene ID
16987
Gene Name
lanosterol synthase
Gene Aliases
2810025N20Rik, D10Ertd116e, Osc
Chromosome Location
Chr.10: 76531565 - 76557139 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK031271.1 - 4 - 5 475 76
AK044016.1 - 4 - 5 469 76
AK054393.1 - 4 - 5 487 76
AK078023.1 - 4 - 5 468 76
AK141216.1 - 4 - 5 487 76
AK154520.1 - 4 - 5 481 76
BC029082.1 - 4 - 5 465 76

View Full Product Details