Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc19a1
- Entrez Gene ID
-
20509
- Gene Name
- solute carrier family 19 (folate transporter), member 1
- Gene Aliases
-
AI323572, RFC, RFC-1, RFC1
- Chromosome Location
-
Chr.10: 77032269 - 77050432 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK050714.1 |
- |
1 - 2 |
165 |
82 |
View Full Product Details