Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap10-4
Entrez Gene ID
100191037
Gene Name
keratin associated protein 10-4
Gene Aliases
675238, A030009A09Rik, AA589524
Chromosome Location
Chr.10: 77826169 - 77827070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135991.1 NP_001129463.1 1 - 1 723 68
GenBank mRNA AK020700.1 - 1 - 1 751 68
BC115499.1 - 1 - 1 723 68
BC115500.1 - 1 - 1 723 68

View Full Product Details