Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gstt4
- Entrez Gene ID
-
75886
- Gene Name
- glutathione S-transferase, theta 4
- Gene Aliases
-
4930583C14Rik, AI507011
- Chromosome Location
-
Chr.10: 75814943 - 75822563 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC052144.1 |
- |
1 - 2 |
219 |
76 |
View Full Product Details