Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmod2
Entrez Gene ID
50876
Gene Name
tropomodulin 2
Gene Aliases
N-Tmod, NTMOD
Chromosome Location
Chr.9: 75565621 - 75611348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF237629.1 - 1 - 2 64 57

View Full Product Details