Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Robo2
Entrez Gene ID
268902
Gene Name
roundabout homolog 2 (Drosophila)
Gene Aliases
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568
Chromosome Location
Chr.16: 73892061 - 75447298 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175549.4 NP_780758.3 9 - 10 1433 81
GenBank mRNA AK035110.1 - 10 - 11 2319 81
AK085461.1 - 10 - 11 2070 81
BC055333.1 - 10 - 11 1753 81
DQ533875.1 - 11 - 12 1725 81
DQ533876.1 - 9 - 10 1433 81

View Full Product Details