Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Raver1
- Entrez Gene ID
-
71766
- Gene Name
- ribonucleoprotein, PTB-binding 1
- Gene Aliases
-
1300006N24Rik, AA050359, mKIAA1978
- Chromosome Location
-
Chr.9: 21074158 - 21091988 on Build GRCm38
- Gene Symbol
- Gm38431
- Entrez Gene ID
-
100568458
- Gene Name
- predicted gene, 38431
- Gene Aliases
-
Raver1-fdx1l
- Chromosome Location
-
Chr.9: 21067514 - 21092008 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038081.2 |
|
10 - 11 |
1905 |
77 |
GenBank mRNA |
AK004919.1 |
- |
10 - 11 |
1898 |
77 |
AY275472.1 |
- |
10 - 11 |
1884 |
77 |
View Full Product Details