The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031196.3 | NP_112473.1 | 1 - 2 | 135 | 79 |
GenBank mRNA | AK079592.1 | - | 1 - 2 | 96 | 79 |
AK085024.2 | - | 1 - 2 | 81 | 79 | |
AK088803.1 | - | 1 - 2 | 98 | 79 | |
BY791593.1 | - | 1 - 2 | 136 | 79 | |
U32469.1 | - | 1 - 2 | 75 | 79 | |
U66103.1 | - | 1 - 2 | 88 | 79 |