Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx22
Entrez Gene ID
382083
Gene Name
sorting nexin 22
Gene Aliases
Gm1120
Chromosome Location
Chr.9: 66065176 - 66069780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025612.2 NP_001020783.1 2 - 3 214 70
GenBank mRNA AK032028.1 - 2 - 3 203 70
BC138950.1 - 2 - 3 201 70
BC138951.1 - 2 - 3 201 70

View Full Product Details