Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnrnpul1
Entrez Gene ID
232989
Gene Name
heterogeneous nuclear ribonucleoprotein U-like 1
Gene Aliases
E130317O14Rik, E1B-AP5, E1BAP5, Hnrnpul, Hnrpul1
Chromosome Location
Chr.7: 25721165 - 25756245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144922.2 NP_659171.1 11 - 12 1772 77
GenBank mRNA AK087545.1 - 11 - 12 1742 77
AK155604.1 - 11 - 12 1783 77
BC021506.1 - 11 - 12 1796 77
BC027844.1 - 8 - 9 1065 77
BC078642.1 - 11 - 12 1696 77

View Full Product Details