The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013373.2 | NP_001013391.2 | 7 - 8 | 921 | 60 |
GenBank mRNA | BC042878.1 | - | 6 - 7 | 657 | 60 |
BC085323.1 | - | 7 - 8 | 906 | 60 |