Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cln6
Entrez Gene ID
76524
Gene Name
ceroid-lipofuscinosis, neuronal 6
Gene Aliases
1810065L06Rik, AW743417, D9Bwg1455e, nclf
Chromosome Location
Chr.9: 62838787 - 62852002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033175.2 NP_001028347.1 1 - 2 166 110
GenBank mRNA AK154414.1 - 1 - 2 167 110

View Full Product Details