Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrna5
Entrez Gene ID
110835
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 5
Gene Aliases
Acra-5, Acra5
Chromosome Location
Chr.9: 54979828 - 55009231 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176844.4 NP_789814.2 4 - 5 543 58
GenBank mRNA AF204689.1 - 3 - 4 308 58
AY574264.1 - 4 - 5 326 58
BC106839.2 - 4 - 5 326 58
BC106853.2 - 4 - 5 326 58
DQ318788.1 - 4 - 5 496 58

View Full Product Details