Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfap2e
Entrez Gene ID
332937
Gene Name
transcription factor AP-2, epsilon
Gene Aliases
AI505200, AP-2e, Tcfap2e
Chromosome Location
Chr.4: 126716003 - 126736269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198960.2 NP_945198.2 1 - 2 304 136
GenBank mRNA AY325902.1 - 1 - 2 144 136
CO809755.1 - 1 - 2 341 136

View Full Product Details