The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270435.1 | NP_001257364.1 | 14 - 15 | 2166 | 96 |
NM_001270446.1 | NP_001257375.1 | 9 - 10 | 1422 | 96 | |
NM_028506.1 | NP_082782.1 | 10 - 11 | 1429 | 96 | |
NM_029286.2 | NP_083562.1 | 14 - 15 | 2181 | 96 | |
GenBank mRNA | AK049010.1 | - | 9 - 10 | 1322 | 96 |
AK078362.1 | - | 14 - 15 | 2108 | 96 | |
AK163185.1 | - | 14 - 15 | 2107 | 96 | |
AK224940.1 | - | 10 - 11 | 1358 | 96 | |
AK224961.1 | - | 9 - 10 | 1421 | 96 | |
BC114436.1 | - | 14 - 15 | 2108 | 96 |