Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smek2
Entrez Gene ID
104570
Gene Name
SMEK homolog 2, suppressor of mek1 (Dictyostelium)
Gene Aliases
AW011752, AW557776, mKIAA1387
Chromosome Location
Chr.11: 29167939 - 29220797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134034.2 NP_598795.2 2 - 3 597 145
GenBank mRNA AK037253.1 - 2 - 3 529 145
AK041513.1 - 2 - 3 542 145
AK090014.1 - 2 - 3 573 145
AK129344.1 - 2 - 3 508 145
AK165940.1 - 2 - 3 521 145
AK166627.1 - 2 - 3 523 145
BC006870.1 - 2 - 3 508 145
CD350450.1 - 2 - 3 597 145

View Full Product Details