The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172495.5 | NP_766083.3 | 10 - 11 | 2434 | 70 |
GenBank mRNA | AK128935.1 | - | 9 - 10 | 2257 | 70 |
BC042791.1 | - | - | 514 | 70 | |
BC076623.1 | - | 8 - 9 | 2040 | 70 | |
BC141150.1 | - | 9 - 10 | 2135 | 70 | |
BC145459.1 | - | 8 - 9 | 2020 | 70 |