Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncoa7
Entrez Gene ID
211329
Gene Name
nuclear receptor coactivator 7
Gene Aliases
9030406N13Rik
Chromosome Location
Chr.10: 30645582 - 30805977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111267.2 NP_001104737.1 2 - 3 363 60
NM_172495.5 NP_766083.3 12 - 13 2714 60
GenBank mRNA AK033503.1 - 2 - 3 277 60
AK042369.1 - 2 - 3 333 60
AK128935.1 - 11 - 12 2537 60
AK165098.1 - 2 - 3 332 60
BC042791.1 - - 794 60
BC049997.1 - 2 - 3 313 60
BC058572.1 - 2 - 3 285 60
BC076623.1 - 10 - 11 2320 60
BC080670.1 - - 287 60
BC141150.1 - 11 - 12 2415 60
BC145459.1 - 10 - 11 2300 60
EF602028.1 - 2 - 3 363 60

View Full Product Details