Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Camk2b
Entrez Gene ID
12323
Gene Name
calcium/calmodulin-dependent protein kinase II, beta
Gene Aliases
-
Chromosome Location
Chr.11: 5969642 - 6065748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174053.1 NP_001167524.1 18 - 19 1620 67
NM_001174054.1 NP_001167525.1 19 - 20 1572 67
NM_007595.5 NP_031621.3 19 - 20 1611 67
GenBank mRNA AK046454.1 - 19 - 20 1612 67
AK158794.1 - 14 - 15 1255 67
AK163698.1 - 19 - 20 1428 67
BC024803.1 - 19 - 20 1388 67
BC080273.1 - 18 - 19 1422 67
X63615.1 - 19 - 20 1426 67

View Full Product Details