The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163507.1 | NP_001156979.1 | 4 - 5 | 1151 | 60 |
NM_027613.1 | NP_081889.1 | 4 - 5 | 1151 | 60 | |
GenBank mRNA | AK036724.1 | - | 4 - 5 | 989 | 60 |
AK137556.1 | - | 4 - 5 | 990 | 60 | |
BC137623.1 | - | 4 - 5 | 951 | 60 | |
BC144789.1 | - | 4 - 5 | 951 | 60 |