Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Reg1
Entrez Gene ID
19692
Gene Name
regenerating islet-derived 1
Gene Aliases
-
Chromosome Location
Chr.6: 78420227 - 78428668 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009042.1 NP_033068.1 2 - 3 127 65
GenBank mRNA AK133506.1 - 2 - 3 131 65
AK160452.1 - 2 - 3 128 65
BC028761.1 - 2 - 3 123 65

View Full Product Details