Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk19
Entrez Gene ID
78334
Gene Name
cyclin-dependent kinase 19
Gene Aliases
2700084L06Rik, AI845279, AW228747, CDK11, Cdc2l6, mKIAA1028
Chromosome Location
Chr.10: 40349308 - 40483818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168304.1 NP_001161776.1 10 - 11 1318 121
NM_001291816.1 NP_001278745.1 9 - 10 1242 121
NM_001291817.1 NP_001278746.1 9 - 10 1260 121
NM_198164.3 NP_937807.2 9 - 10 1186 121
GenBank mRNA AK028643.1 - - 887 121
AK052818.1 - 10 - 11 1149 121
AK053651.1 - 9 - 10 1159 121
AK122421.1 - 10 - 11 1318 121
AK147340.1 - 9 - 10 1040 121
BC030312.1 - 3 - 4 214 121
BC048908.1 - 10 - 11 1068 121
BC062100.1 - 9 - 10 1042 121
BC080280.1 - 9 - 10 1192 121
BC137614.1 - 10 - 11 1272 121
BC144782.1 - 10 - 11 1272 121

View Full Product Details