The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168304.1 | NP_001161776.1 | 12 - 13 | 1664 | 60 |
NM_001291816.1 | NP_001278745.1 | 11 - 12 | 1588 | 60 | |
NM_001291817.1 | NP_001278746.1 | 11 - 12 | 1606 | 60 | |
NM_198164.3 | NP_937807.2 | 11 - 12 | 1532 | 60 | |
GenBank mRNA | AK028643.1 | - | - | 1234 | 60 |
AK052818.1 | - | 12 - 13 | 1495 | 60 | |
AK053651.1 | - | 11 - 12 | 1505 | 60 | |
AK122421.1 | - | 12 - 13 | 1664 | 60 | |
AK147340.1 | - | 11 - 12 | 1386 | 60 | |
BC030312.1 | - | 5 - 6 | 560 | 60 | |
BC048908.1 | - | 12 - 13 | 1492 | 60 | |
BC062100.1 | - | 11 - 12 | 1388 | 60 | |
BC080280.1 | - | 11 - 12 | 1538 | 60 | |
BC137614.1 | - | 12 - 13 | 1618 | 60 | |
BC144782.1 | - | 12 - 13 | 1618 | 60 |