Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smyd5
Entrez Gene ID
232187
Gene Name
SET and MYND domain containing 5
Gene Aliases
AW536703, NN8-4AG, Rai15, Rrg1
Chromosome Location
Chr.6: 85431976 - 85446429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144918.2 NP_659167.2 8 - 9 804 63
GenBank mRNA AK158267.1 - 8 - 9 785 63
AK162926.1 - 8 - 9 728 63
BC016525.1 - 8 - 9 791 63

View Full Product Details