The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026743.3 | NP_081019.1 | 1 - 2 | 258 | 78 |
GenBank mRNA | AK003691.1 | - | 1 - 2 | 110 | 78 |
AK028808.1 | - | 1 - 2 | 149 | 78 | |
AK053788.1 | - | 1 - 2 | 245 | 78 | |
AK087294.1 | - | 1 - 2 | 273 | 78 | |
AK090022.1 | - | 1 - 2 | 258 | 78 | |
BC057639.1 | - | 1 - 2 | 100 | 78 |