Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec9a
Entrez Gene ID
232414
Gene Name
C-type lectin domain family 9, member a
Gene Aliases
9830005G06Rik, DNGR-1
Chromosome Location
Chr.6: 129408862 - 129501701 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205363.1 NP_001192292.1 6 - 7 762 85
NM_001205364.1 NP_001192293.1 5 - 6 681 85
NM_172732.3 NP_766320.2 5 - 6 684 85
GenBank mRNA AK041288.1 - 5 - 6 684 85
BC087955.1 - 6 - 7 761 85
EU339277.1 - 5 - 6 591 85
EU339278.1 - 6 - 7 669 85
EU339279.1 - 5 - 6 588 85

View Full Product Details