Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcp1b
Entrez Gene ID
319618
Gene Name
DCP1 decapping enzyme homolog B (S. cerevisiae)
Gene Aliases
B930050E02Rik
Chromosome Location
Chr.6: 119175251 - 119222737 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033379.3 NP_001028551.2 3 - 4 359 92
GenBank mRNA AK049193.1 - 3 - 4 287 92
AK050548.1 - 3 - 4 324 92
AK153860.1 - 3 - 4 361 92
AK216011.1 - 3 - 4 221 92
BC141162.1 - 3 - 4 338 92
BC141163.1 - 3 - 4 338 92

View Full Product Details