The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020257.2 | NP_064653.1 | 2 - 3 | 153 | 73 |
GenBank mRNA | AF121352.1 | - | 2 - 3 | 126 | 73 |
AF350411.1 | - | 2 - 3 | 130 | 73 | |
AY137345.1 | - | 2 - 3 | 152 | 73 | |
BC139192.1 | - | 2 - 3 | 126 | 73 | |
BC139193.1 | - | 2 - 3 | 126 | 73 | |
HQ713440.1 | - | 1 - 2 | 60 | 73 |