The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027526.1 | NP_081802.1 | 7 - 8 | 928 | 56 |
GenBank mRNA | AK018120.1 | - | 6 - 7 | 759 | 56 |
AK158756.1 | - | 8 - 9 | 954 | 56 | |
BC138284.1 | - | 7 - 8 | 894 | 56 | |
BC145232.1 | - | 7 - 8 | 921 | 56 | |
BC145752.1 | - | 7 - 8 | 894 | 56 |