The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164557.1 | NP_001158029.1 | 3 - 4 | 712 | 71 |
NM_001164558.1 | NP_001158030.1 | 4 - 5 | 409 | 71 | |
NM_026018.3 | NP_080294.1 | 4 - 5 | 480 | 71 | |
GenBank mRNA | AB009296.1 | - | 3 - 4 | 290 | 71 |
AK002288.1 | - | 2 - 3 | 417 | 71 | |
AK007936.1 | - | 3 - 4 | 698 | 71 | |
AK008253.1 | - | 4 - 5 | 466 | 71 | |
AK012305.1 | - | 4 - 5 | 427 | 71 | |
BC013542.1 | - | 3 - 4 | 317 | 71 | |
DV057288.1 | - | 4 - 5 | 426 | 71 | |
DV058382.1 | - | 3 - 4 | 401 | 71 |