The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010696.3 | NP_034826.2 | 19 - 20 | 1479 | 82 |
GenBank mRNA | AK036727.1 | - | 19 - 20 | 1537 | 82 |
AK037970.1 | - | 19 - 20 | 1513 | 82 | |
AK088552.1 | - | 19 - 20 | 1601 | 82 | |
AK153374.1 | - | 19 - 20 | 1484 | 82 | |
AK168134.1 | - | 19 - 20 | 1494 | 82 | |
AK170491.1 | - | 19 - 20 | 1756 | 82 | |
BC006948.1 | - | 19 - 20 | 1479 | 82 | |
U20159.1 | - | 19 - 20 | 1518 | 82 |