The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282993.1 | NP_001269922.1 | 7 - 8 | 1332 | 155 |
NM_001282994.1 | NP_001269923.1 | 8 - 9 | 1238 | 155 | |
NM_172496.3 | NP_766084.3 | 8 - 9 | 1407 | 155 | |
GenBank mRNA | AK049730.1 | - | 8 - 9 | 1200 | 155 |
AK122334.1 | - | 8 - 9 | 1238 | 155 | |
AK147366.1 | - | 7 - 8 | 1332 | 155 | |
AK147633.1 | - | 7 - 8 | 1332 | 155 | |
AY308745.1 | - | 8 - 9 | 1247 | 155 | |
BC145118.1 | - | 8 - 9 | 1247 | 155 | |
BC150706.1 | - | 8 - 9 | 1247 | 155 | |
U26967.1 | - | 7 - 8 | 980 | 155 |