Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf19b
Entrez Gene ID
75234
Gene Name
ring finger protein 19B
Gene Aliases
4930534K13Rik, 4930555L03Rik, Ibrdc3
Chromosome Location
Chr.4: 129058174 - 129085886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK089131.1 - 1 - 2 68 58
BC139808.1 - 2 - 3 391 58

View Full Product Details